To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- CAS No.:9003-98-9
- Size:1000 units
- Storage conditions:Freezer
- Enzyme name:Desoxyribonuclease I (DNase I)
- Cat. no.:CA101228-386
Specifications
About this item
DNase I, (RNase-free) is an endonuclease that nonspecifically cleaves DNA to release di-, tri- and oligonucleotide products with 5´-phosphorylated and 3´-hydroxylated ends
DNase I acts on single- and double-stranded DNA, chromatin and RNA:DNA hybrids.