ContactUsLinkComponent
M-MuLV Reverse Transcriptase 10,000 units
M-MuLV Reverse Transcriptase
Catalog #: CA101228-288
CAS Number:  
M-MuLV Reverse Transcriptase 10,000 units
M-MuLV Reverse Transcriptase
Catalog #: CA101228-288
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    10000 units
  • Enzyme name:
    M-MuLV Reverse Transcriptase
  • Cat. no.:
    CA101228-288
Specifications
About this item
Moloney Murine Leukemia Virus (M-MuLV, MMLV) Reverse Transcriptase is an RNA-directed DNA polymerase
This enzyme can synthesize a complementary DNA strand initiating from a primer using either RNA (cDNA synthesis) or single-stranded DNA as a template. M-MuLV Reverse Transcriptase lacks 3´ → 5´ exonuclease activity.
Customers Who Bought This Also Bought