Order Entry
ContactUsLinkComponent
T4 DNA Ligase 100,000 units
T4 DNA Ligase, New England Biolabs
Catalog #: CA101228-176
CAS Number:  
T4 DNA Ligase 100,000 units
T4 DNA Ligase, New England Biolabs
Catalog #: CA101228-176
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    400,000 U/ml
  • Description:
    T4 DNA Ligase
  • Size:
    100000 units
  • Cat. no.:
    CA101228-176

Specifications

About this item

Catalyzes the formation of a phosphodiester bond between juxtaposed 5' phosphate and 3' hydroxyl termini in duplex DNA or RNA.

This enzyme will join blunt end and cohesive end termini as well as repair single stranded nicks in duplex DNA and some DNA/RNA hybrids. T4 DNA ligase will seal nicks for these DNA substrates.

Customers Who Bought This Also Bought