Order Entry
ContactUsLinkComponent
EZFlow* Syringe Filter, CA, Pore Size: 0.45um, Diameter: 33mm, Sterile
EZFlow® Syringe Filter, CA, Sterile, Foxx Life Sciences
Catalog #: 76018-888
Supplier:  Foxx Life Sciences
EZFlow* Syringe Filter, CA, Pore Size: 0.45um, Diameter: 33mm, Sterile
EZFlow® Syringe Filter, CA, Sterile, Foxx Life Sciences
Catalog #: 76018-888
Supplier:  Foxx Life Sciences

Specifications

  • Ø:
    33 mm
  • Pore size:
    0.45 µm
  • Sterility:
    Sterile
  • Cat. no.:
    76018-888

Specifications

About this item

CA syringe filters are low protein binding, have good flow rates, and are great for use with media and buffers.

  • Low protein binding
  • Good flow rates
  • Great for use with medica, buffers

EZFlow® 13, 25, and 33 mm syringe filters are used for prefiltration, clarification, sterilization, or fine particulate removal. These syringe filters are available in a variety of filter membrane materials in both 0.2 and 0.45 µm pore sizes. The filter membranes are reinforced with polypropylene housing, with a female Luer-lok inlet and a male Luer slip outlet. All materials comply with USP Class VI Standards and are available for both sterile and non-sterile applications.

Certifications: All materials comply with USP Class VI Standards and FDA compliances.
Customers Who Bought This Also Bought