Order Entry
ContactUsLinkComponent
EZFlow* Syringe Filter 25mm 0.22um PES
EZFlow® High Particulate Syringe Filter, PES with Glass Fiber Prefilter, Foxx Life Sciences
Catalog #: 76018-878
Supplier:  Foxx Life Sciences
EZFlow* Syringe Filter 25mm 0.22um PES
EZFlow® High Particulate Syringe Filter, PES with Glass Fiber Prefilter, Foxx Life Sciences
Catalog #: 76018-878
Supplier:  Foxx Life Sciences

Specifications

  • Ø:
    25 mm
  • Pore size:
    0.22 µm
  • Cat. no.:
    76018-878
  • Membrane:
    Glass Fiber and PES

Specifications

About this item

High particulate PES syringe filters have a 1.0 µm glass fiber prefilter designed for high throughputs and flow rates.

  • Super high flow rates
  • Very low protein binding
  • Very low extractables
  • Perfect for use with media, buffers, and DNA

EZFlow® 25 mm high particulate syringe filters are designed for high throughput filtration of viscous and particle-laden solutions. The syringe filter contains a filter membrane along with a glass fiber prefilter. The filter membrane is available in PES or Hydrophilic PVDF materials, in 0.2 or 0.45 µm pore sizes. The filter membrane is reinforced with a polypropylene housing with a female Luer-Lok inlet and a male Luer Slip outlet.

They also have very low extractables.

Certifications: All materials comply with USP Class VI Standards and FDA compliances.
Customers Who Bought This Also Bought