EZFlow® Syringe Filter, PES, Sterile, Foxx Life Sciences
Catalog #: 76018-866
Supplier: Foxx Life Sciences
Specifications
- Ø:13 mm
- Pore size:0.22 µm
- Sterility:Sterile
- Cat. no.:76018-866
Specifications
About this item
High performance PES syringe filters have super high flow rates, very low extractables, and are very low protein binding.
- Super high flow rates
- Very low protein binding
- Very low extractables
- Perfect for use with media, buffers, and DNA
EZFlow® 13, 25, and 33 mm Syringe Filters are used for prefiltration, clarification, sterilization, or fine particulate removal. These syringe filters are available in a variety of filter membrane materials in both 0.2 and 0.45 µm pore sizes. The filter membranes are reinforced with polypropylene housing, with a female Luer-lok inlet and a male Luer slip outlet.
All at an affordable cost.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.
Customers Who Bought This Also Bought