EZFlow® Membrane Disc Filter, PES, Foxx Life Sciences
Catalog #: 76018-800
Supplier: Foxx Life Sciences
Specifications
- Ø:47 mm
- Pore size:0.45 µm
- Sterility:Non sterile
Specifications
About this item
High performance PES disc filters have super high flow rates.
- Very low protein binding
- Very low extractables
- Perfect for use with media, buffers, and DNA
EZFlow® 13, 25, 47 and 90 mm membrane disc filters are used to retain particles or microorganisms by surface capture. The membrane is offered in 0.22 and 0.45 µm pore sizes.
All at an affordable cost.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.
Customers Who Bought This Also Bought