EZBio® Single-Use Assembly, Media Bottle, PETG, Vented with Tubing, Sterile, Foxx Life Sciences
Catalog #: 76018-354
Supplier: Foxx Life Sciences
Specifications
- Capacity:1000 ml
- Sterility:Sterile
- Material:PETG
- Cat. no.:76018-354
Specifications
About this item
EZBio® single-use PETG bottle assemblies are an innovative and cost efficient option for single-use applications.
- Perfect for fluid transfer and storage
- VersaCap® technology
- More cost efficient than stainless steel apparatuses
- BPA-free
- Sterilized
The EZBio® single-use sterile media bottles with tubing and vent vilters are perfect for fluid transfer and storage. Assemblies are made entirely with USP Class VI components and Gamma Irradiated in the United States. Assemblies take advantage of Foxx’s VersaCap® technology, which means there is no machining necessary to produce the product. Perfect for single-use applications and to phase out costly stainless steel apparatuses that are expensive to clean and validate.
The caps for these bottles are vented with tubing, and sterilized.
Certifications: All materials comply with USP Class VI standards.
Customers Who Bought This Also Bought