Magnetic Separation Rack, 6-Tube, New England Biolabs
Catalog #: CA101227-252
Supplier: New England Biolabs (NEB)
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:6-Tube Magnetic Separation Rack
- Cat. no.:CA101227-252
Specifications
About this item
The Magnetic Separation Racks are designed to be used for small-scale separations using magnetic particles
The magnets are located on the sides of the racks, resulting in minimal sample loss during pipetting.
Customers Who Bought This Also Bought