UV/VIS Photometric Accuracy Calibration Standards, FUV Dual Series, FireFlySci
Catalog #: 76016-732
Supplier: FIREFLYSCI, INC.
CAS Number:
Specifications
- Description:Photometric accuracy & stray light calibration standard
- Size:12.5 x 12.5 x 45 mm
- Cat. no.:76016-732
- Density:0.5 au
- Accuracy:±0.0022 au
Specifications
About this item
Firefly’s FUV Dual Series filters allow you to verify photometric accuracy and stray light in the range of 200 - 700 nm. Includes certified values for both the front and back of the filter for determining stray light.
- Highly accurate and consistent spectrophotometer calibration every time
- Never has to be replaced
- Never has to be recalibrated if used properly
- Scratch-resistant coating protects optical surfaces
- Can be used in any spectrophotometer (with standard 12.5 x 12.5 x 45mm holder)
- Surpasses NIST-2031 requirements and tolerances
Certifications: Certificate of calibration with NIST traceability.
Delivery: Supplied with instruction manual.
Caution: Note: The FUV-0.2D and below are highly sensitive and only for use with customers with the following high-end machines: Cary-5000, Cary-7000, Lambda 900, Lambda 950, Lambda 1050 or Shimadzu UV-3600.
Customers Who Bought This Also Bought