U-Demountable Cuvettes with Removable Windows, FireFlySci
Catalog #: 76015-622
Supplier: FIREFLYSCI, INC.
Specifications
- Capacity:0.020 ml
- Lightpath:0.05 mm
- Cat. no.:76015-622
Specifications
About this item
U-demountable cuvettes have two polished windows.
Ordering information: A20 cell mount not included.
Customers Who Bought This Also Bought