U-Demountable Cuvettes with Removable Windows, FireFlySci
Catalog #: 76015-618
Supplier: FIREFLYSCI, INC.
Specifications
- Capacity:0.400 ml
- Lightpath:1 mm
- Cat. no.:76015-618
Specifications
About this item
U-demountable cuvettes have two polished windows.
Ordering information: A20 cell mount not included.
Customers Who Bought This Also Bought