VWR® Tube Top Vacuum Filters
Catalog #: 76012-774
Supplier: VWR International
Specifications
- Pore size:0.20 µm
- Description:150 mL Upper Cup Capacity
Specifications
About this item
Minimize unnecessary transfers by filtering directly into 50 ml centrifuge tubes.
- Tube top vacuum filters
- 50 mm Ø membrane
- Two pore sizes: 0.2 and 0.45 μm
- Gamma radiation sterilized
- Non-pyrogenic
PES membranes offer low protein binding and low extractables.
The 0.2 µm pore size is ideal for sterilization applications and media preparation. The 0.45 µm pore size is ideal for buffer clarification.
Ideal for tissue culture applications.
Certifications: ISO13485 :2003 and ISO9001:2008.
BodyTextAccessories: The Tube Top Filter Stand features 7 openings, suitable for holding 1.5 and 2.0 ml microcentrifuge tubes, or 15 and 50 ml conical centrifuge tubes. It provides an operating temperature range of –80 to 121 °C, and may be autoclaved.
Delivery: Each polypropylene centrifuge tube is supplied with an individually wrapped cap for storage.
Packaging: Packaged in resealable bags. Each individual unit is lot-numbered for easy identification and tracking.
Customers Who Bought This Also Bought