VWR® Nonsterile Safety Goggles
Catalog #: 76017-902
Supplier: VWR International
Specifications
- Lens Coating:Uncoated
- Description:Indirect ventilation
- Frame Material:PVC
- Lens Material:PC
Specifications
About this item
Economical non sterile safety goggles are designed to protect from hazards with a flexible ridge around a PVC frame.
- Two ventilation options available
- Goggles fit over most prescription glasses
- Fully adjustable headband
- Provides 99.9% UV protection
These economical, lightweight, flexible mask goggles offer unmatched comfort and protection.
The indirect vent goggles feature quad vents to minimize lens fogging. Indirect vents offer protection from chemical splashes and impact.
The direct vent goggles offer direct perforations in the mask to provide maximum airflow and prevent fogging. Direct vent option provides protection from flying particles.
They can be worn over personal glasses. A broad bearing surface around the edge of the mask molds snugly to the wearer's face. The anti-fog lens coating provides superior fogging protection. The goggles meet ANSI Z87 requirements.
Certifications: Meet ANSI Z87+ 1-2015 standards.
Customers Who Bought This Also Bought