White Polypropylene Screw Caps, F217 & Fluoropolymer Resin Liner, Qorpak®
Catalog #: 76012-836
Supplier: Qorpak
Specifications
- Cap Size:28-400
- Cat. no.:76012-836
Specifications
About this item
Polypropylene caps with F217 & Fluoropolymer Resin liners are an economical alternative to Thermoset & Phenolic Fluoropolymer Resin lined caps.
- Liners are very resilient – great sealing capability
- Liners have strong resistance to chemicals and moisture
Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Customers Who Bought This Also Bought