VWR® Spinbar® Micro Stir Bars
Catalog #: 76001-872
Supplier: VWR International
Specifications
- Coating:PTFE
- Ø:2 mm (1/16")
- Length:2 mm (1/16")
- Description:Stir Bar
- Cat. no.:76001-872
Specifications
About this item
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes, and gradient makers.
- Particularly useful for environmental testing
- Valuable for preparing and evaluating small sample volumes
- Teflon® PTFE encapsulated
- Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Customers Who Bought This Also Bought