Locking Microcentrifuge Tubes, Globe Scientific
Catalog #: 76004-128
Supplier: Globe Scientific
Specifications
- Capacity:2.0 ml
- Color:Natural
- Cat. no.:76004-128
- Packaging:Bag/500
Specifications
About this item
These tubes are engineered with a secure locking system that provides the security of a screw cap microtube with the simplicity of a snap cap tube.
- Maintain sample integrity with locking cap which prevents accidental cap opening and evaporation during contrifugation
- Single-use tubes for laboratory procedures including centrifugation, boiling, incubation, freezing, and sample storage
- Tubes are made from optimally clear USP Class VI medical grade polypropylene
- Tubes feature graduations, writing area, piercable cap, are autoclavable, and are packaged in zip lock, re-closable bags
Manufactured in a class 100000 cleanroom and certified to be free from RNase and DNase, Human DNA, Pyrogens, PCR Inhibitors, and ATP.
Customers Who Bought This Also Bought