Slimline™ Flathead Mop Covers and Hardware, Micronova
Catalog #: CA76006-322
Supplier: Micronova
Specifications
- Description:Microfiber single ply slipcover
- Material:Blue microfiber 80% polyester or 20% polyamide (without foam)
- Cat. no.:CA76006-322
- Width:36 cm (14 ")
Specifications
About this item
A series of elasticized slipcovers, with urethane foam interiors, that are ideal for fine cleaning and sanitizing all surfaces and equipment in the cleanroom. Available without a foam interior or gamma irradiated for aseptic environments.
- Covers are easy to change and fit securely over mop frame
- Fully encased Urethane Foam layer for ample disinfectant dispersal
- Single-ply covers available without foam interior
- Fits on QPSL plastic frame or QDSL stainless steel frame
- Suitable for use with strong disinfectants and repeated autoclaving
The Micronova Slimline™ Flat Head Mop Covers fit easily and securely onto the 360° swivel SlimLine™ plastic or stainless steel mop adapters, which can be locked at 180° for more controlled cleaning.
Customers Who Bought This Also Bought