KIMTECH PURE® A8 Cleanroom Coverall, Kimberly-Clark
Catalog #: CA75832-284
Supplier: Kimberly-Clark
Specifications
- Clothing Size:M
- Garment application:Cleanroom,Laboratory
- Description:KIMTECH PURE® A8 Cleanroom Coverall
- Environmentally Preferable:Waste Reducing
- Cat. no.:CA75832-284
Specifications
About this item
KIMTECH PURE® A8 Cleanroom Coveralls are designed for quality, comfort, and enhanced productivity.
- Lightweight, innovative fabric moves with the worker
- Built-in thumb loops to protect people and processes
- High neck collar to control contamination risk
- Recyclable through RightCycle by Kimberly-Clark Professional®
The most critical environments require a garment that will enable workers to do their very best
Applications include:
ISO 7/8 Grade C/D
Nonsterile Pharmaceutical Manufacturing
Bio-Medical Research
Pharmaceutical Compounding
Biotechnology
One that provides protection as well as comfort, because more comfortable garments can increase productivity and reduce human errors by 56%. Protect workers and the quality of their work with the KIMTECH PURE® A8 Cleanroom Coverall.
Packaging: One large bag of 25 bulk packaged coveralls per case.
Customers Who Bought This Also Bought