VWR® Pouring Ring for Bottles
Catalog #: 75983-266
Supplier: VWR International
Specifications
- Color:White
- Description:Pouring Ring, GL 45
- Cat. no.:75983-266
- For Use With:Bottles
Specifications
About this item
Made from polyethylene.
- Autoclavable up to 140 °C
- White color
For GL45 storage bottle.
Customers Who Bought This Also Bought