Specifications
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:Mag-Bind® TotalPure NGS
- Description:Mag-Bind® TotalPure NGS
- Cat. no.:CA75877-714
- Volume:5 ml
Specifications
About this item
The Mag-Bind® TotalPure NGS Kit offers an easy-to-use, reliable solution for purification of both DNA and RNA for next-generation sequencing workflows with high recovery rates.
Mag-bind TotalPure NGS is capable of selectively binding fragments depending on the reagent to sample ratio used giving the user flexibility to perform left, right or double-sided size selection. This kit is designed for both manual and fully automated purification of DNA, RNA samples as well as for the purification of PCR products. The system combines Omega Bio-tek’s proprietary chemistries with reversible nucleic acid-binding properties of magnetic beads to selectively bind fragments larger than 100 bp and eliminates excess nucleotides, primers and small, non-targeted products such as primer-dimers. Purified DNA and RNA is suitable for a variety of downstream applications such as NGS library preparation, microarrays, automated fluorescent sequencing, and restriction enzyme digestion.