To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Mitochondrial Marker
- Clonality:Monoclonal
- Clone:113-1
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Conjugation:Biotin
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 μL
- Storage buffer:PBS/0.1% BSA/0.05% azide
- Concentration:0.1 mg/mL
- Cat. no.:75961-662
- Supplier No.:BNCB0739-100
Specifications
About this item
This MAb recognizes a 60 kDa antigen associated with the mitochondria in human cells. It can be used to stain mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignant cells. This antibody is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells, including neurons and embryonic stem cells. Immunostaining pattern with anti-mitochondrial MAb has been reported as a useful discriminatory adjunct in the complex differential diagnosis of granular renal cell tumors. Reportedly, this MAb facilitates the classification of salivary tumors.
Type: Primary
Antigen: Mitochondrial Marker
Clonality: Monoclonal
Clone: Clone 113-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human