Order Entry
ContactUsLinkComponent
QuantiChrom* TBARS Assay Kit 100 tests
QuantiChrom™ TBARS (Lipid Peroxide) Assay Kit, BioAssay Systems
Catalog #: 75878-012
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
QuantiChrom* TBARS Assay Kit 100 tests
QuantiChrom™ TBARS (Lipid Peroxide) Assay Kit, BioAssay Systems
Catalog #: 75878-012
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    TBARS (Lipid Peroxide) Assay Kit
  • Size:
    100 assays
  • Method:
    OD535 nm, or FL560/585 nm
  • Sample:
    Serum, plasma, urine, saliva etc
  • Processing time:
    80 minutes
  • Cat. no.:
    75878-012
  • Supplier No.:
    DTBA-100

Specifications

About this item

For quantitative determination of lipid peroxides (thiobarbituric acid reactive substances, TBARS).

  • Linear detection range colorimetric assay 1 - 30 μM and fluorometric assay 0.1 - 1.5 μM MDA

Oxidative attack of essential cell components by reactive oxygen species has been associated with several human diseases, such as atherosclerosis, cardiovascular diseases, diabetes, liver disorders, and inflammatory rheumatic diseases

BioAssay Systems’ TBARS assay is based on the reaction of TBARS with thiobarbituric acid (TBA) to form a pink colored product. The color intensity at 535nm or fluorescence intensity at (λex/em = 560nm/585nm) is directly proportional to TBARS concentration in the sample.

THIOBARBITURIC ACID REACTIVE SUBSTANCES (TBARS) are low-molecular-weight end products (mainly malondialdehyde, MDA) that are formed during the decomposition of lipid peroxidation products. Increased levels of TBARS have been demonstrated in these diseases. Simple, direct and accurate assays for TBARS find wide applications in research and drug discovery.

Customers Who Bought This Also Bought