ContactUsLinkComponent
QuantiChrom* Creatinine Assay Kit 500 tests
QuantiChrom™ Creatinine Assay Kit, BioAssay Systems
Catalog #: 75877-990
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
QuantiChrom* Creatinine Assay Kit 500 tests
QuantiChrom™ Creatinine Assay Kit, BioAssay Systems
Catalog #: 75877-990
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Creatinine Assay Kit
  • Size:
    500 assays
  • Method:
    OD510 nm
  • Sample:
    Urine, serum, plasma and biological preparations.
  • Processing time:
    20 minutes
  • Cat. no.:
    75877-990
  • Supplier No.:
    DICT-500
Specifications
About this item
For quantitative determination of creatinine and evaluation of drug effects on creatinine metabolism.
  • Sensitive
  • Detection limit of 0.10 mg/dL (8 μM) creatinine
  • Can be automated as a high-throughput assay for thousands of samples per day
Creatinine is synthesized in the body at a fairly constant rate from creatine, which is produced during muscle contractions from creatine phosphate
BioAssay Systems’ creatinine assay kit is designed to measure creatinine directly in biological samples without any pretreatment. The improved Jaffe method utilizes picrate that forms a red colored complex with creatinine. The intensity of the color, measured at 510nm, is directly proportional to creatinine concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw sample.
In the blood, creatinine is removed by filtration through the glomeruli of the kidney and is secreted into urine. In healthy individuals, creatinine secretion is independent of diet and is fairly constant. The creatinine clearance test has become one of the most sensitive tests for measuring glomerular filtration rate. In kidney disease, creatinine levels in the blood are elevated, whereas the creatinine clearance rate and hence the urine levels are diminished. Creatinine test is most widely used to assess kidney function. Simple, direct and automation-ready. Procedures for measuring creatinine concentration in biological samples are becoming popular in Research and Drug Discovery.
Customers Who Bought This Also Bought