Specifications
- Clothing Size:S/M
- Garment application:Medical,Clinical,Pharmaceutical
- Closure type:Back fastening
- Color:Blue
- Environmentally Preferable:Waste Reducing
- Cat. no.:CA75870-726
- Supplier No.:47992
Specifications
About this item
When working with blood-borne pathogens, biologics, or other harmful liquids, workers must seek advanced protection against spills and splashes. The Kimtech™ A7 certified liquid barrier gown is an ideal solution.
- Bulk packaged
- Thumb loops to maintain glove and garment interface
- Seamless front and closed back
- Easy to don and doff design
Kimtech™ A7 certified liquid barrier gowns deliver stringent barrier protection for people and superior contamination control for any facility. This protective gown has plenty of innovative features, including a seamless front design and an easy to don-and-doff fit to reduce contamination risk and prevent hazardous liquids from getting on clothing or skin. The material offers enhanced liquid barrier protection fabric tested for blood and blood-borne pathogen penetration, chemotherapy drug, and hormone permeation (passes ASTM F1670 for penetration of blood and body fluids and ASTM F1671 for blood borne pathogens).
The built-in thumb loops help maintain glove and garment interface. The gowns are low lint to minimize contamination. Kimtech™ A7 Certified liquid barrier gowns are bulk-packaged and available in a range of sizes to help ensure comfort and compliance.