ENCORE® Perry Style 42®, Surgical Gloves, Ansell
Catalog #: 75874-010
Supplier: Ansell Healthcare
Specifications
- Glove Color:White
- Glove size:6,5
- Powdered/Powder-free:Powder-free
- Latex/Latex-free:Latex
- Finish:Smooth
- Cat. no.:75874-010
- Supplier No.:5711102PF
Specifications
About this item
Latex, powder-free surgical glove with a smooth finish and strong formulation for everyday protection.
- Comfort and fit to help reduce hand fatigue
- Polymer coated for fast and easy donning
- Beaded cuff to help prevent cuff roll down
Customers Who Bought This Also Bought