VWR® Erlenmeyer Flasks, Narrow Mouth
Catalog #: 75870-512
Supplier: VWR International
Specifications
- Capacity:25 / 50 / 125 / 250 / 300 ml
- Description:Erlenmeyer flask sample kit
- Cat. no.:75870-512
Specifications
About this item
Flasks are durable and come with reinforced rim for easy pouring and also avoids any breakage while washing.
- Borosilicate glass fabrication
- Durable design
Feature durable white ceramic enamel scale which indicate approximate volume for measuring and mixing.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1404 Type 1.
Customers Who Bought This Also Bought