COOLING BATH CC-K6 110V/50/60HZ 1.19 GAL
Catalog #: MP10221-018
Supplier: Huber
Customers Who Bought This Also Bought
Hydrion Insta-Chek<sup>®</sup> pH Paper, Micro Essential Laboratory
Supplier: Micro Essential
$14.35 to $142.61
Tetrachloroethylene ≥99.8%, ULTRA RESI-ANALYZED® for organic trace analysis, for IR spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$1,329.84 to $3,816.49
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
$131.08 to $405.43
Silica gel, granulated 1 - 3 mm drying agent, with moisture indicator (orange gel), Supelco®
Supplier: MilliporeSigma
$269.66 to $1,471.88
SGE Syringes, General Purpose Manual Syringe, PTFE Tipped Plunger, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
$219.34