Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:Blue,Green
- Glove size:8,5 - 9
- Length:30 cm (11 13/16")
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Disposable
- Thickness:0.198 mm (7.8 mil)
- Cuff style:Beaded
- Cat. no.:CA75832-940
- Supplier No.:93260090
Specifications
About this item
Disposable chemical resistant gloves.
- Three layer design for superior protection against harsh chemicals including acids, solvents, and bases
- Thin mil construction provides enhanced tactility and dexterity
- Extra-soft material and ergonomic design provide outstanding fit, feel, and flexibility for longer wear time
- Lower acceptable pinhole rate (0.65 AQL) and extended cuff for reliable protection against hazardous substances
- Silicone-free formulation and processing ensure better product protection
- For single use only
The Microflex® 93-260 series glove's unique, triple-layer construction features a soft interior specifically designed to enable easy donning and doffing.
Microflex® 93-260 Gloves are suitable for applications in Blending, compounding materials; Handling aerospace equipment and parts; Handling of painting tools including spray guns and robots; Handling unexpected leaks, spills, or other releases; Maintenance and equipment clean up; Mounting and dismantling, assembly; Petrochemicals processing; Routine and experimental testing; Sample taking and lab processing; Transferring liquid and solids.
A flexible middle layer delivers maximum protection against acids and bases, while the durable exterior delivers maximum protection against organic solvents. Because the glove's tough exterior stands up longer to demanding physical and chemical conditions, workers may change gloves less frequently, resulting in enhanced productivity.