EPP WO Vial and Screw cap RSA-Pro
Catalog #: 75846-062
Supplier: MicroSolv Technology Corp
Customers Who Bought This Also Bought
VRB (Violet Red Bile) Agar, Fluorocult®, Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
1 692,83 $
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) 99%
Supplier: Thermo Scientific Chemicals
57,01 $
Tetrahydrofuran ≥99% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
690,06 $ to 2 135,25 $
Fluoride Standard, 1 ml = 0.1 mg F-, 100 ppm F-, Ricca Chemical Company
Supplier: Ricca Chemical
162,76 $
Micromesh™ Biopsy Cassettes in Quickload™ Sleeves, Simport Scientific
Supplier: Simport Scientific
942,84 $
Chromium(III) oxide ≥99.97% (metals basis), powder, Puratronic® -22 Mesh
Supplier: Thermo Scientific Chemicals
265,99 $