Order Entry
ContactUsLinkComponent
Foam Brick Polar Pack 22 Oz
PolarPack® Water Based Foam Bricks, Sonoco ThermoSafe
Catalog #: 75845-344
Supplier:  Sonoco Thermosafe
Foam Brick Polar Pack 22 Oz
PolarPack® Water Based Foam Bricks, Sonoco ThermoSafe
Catalog #: 75845-344
Supplier:  Sonoco Thermosafe

Specifications

  • Cat. no.:
    75845-344
  • L×W×D:
    22.8×12.1×2.5 cm(9×4.80×1")
  • Weight:
    624 g (22 oz.)
  • Supplier No.:
    FPP22

Specifications

About this item

These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.

  • Vacuum-sealed for consistent weight and shape
  • Perfect when you need maximum surface contact to your payload
  • Available in multiple sizes to fit your packing configuration

These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.

Suitable for products that need refrigerated or frozen protection.

Customers Who Bought This Also Bought