Energy Stick
Catalog #: 470223-984
Supplier: Be Amazing! Toys
Specifications
- Description:Energy Stick
- Size:7 x 1.5"
- Cat. no.:470223-984
- Supplier No.:OHM-350
Specifications
About this item
Energize your lessons on electricity.
- Demonstrate open and closed circuits
- No danger of shock
- Colored lights
Safely demonstrate open and closed series circuits without any danger of electric shocks! When the tube is 'turned on,' the red, green, and blue lights light up and it makes a funny buzzing sound. Activating the tube is the fun part - just touch the metal ends with both hands. Your body forms the conducting material that bridges the gap between the two metal strips and closes the circuit!
Customers Who Bought This Also Bought