Order Entry
ContactUsLinkComponent
Fast-Freeze Flask 600ml
Fast-Freeze® Flasks and Adapters, Labconco®
Catalog #: 75841-040
Supplier:  Labconco
Fast-Freeze Flask 600ml
Fast-Freeze® Flasks and Adapters, Labconco®
Catalog #: 75841-040
Supplier:  Labconco

Specifications

  • Description:
    Fast-Freeze Flask, 600 mL, Amber
  • Cat. no.:
    75841-040
  • Neck Diameter:
    19 mm (¾")
  • Supplier No.:
    7540801

Specifications

About this item

Fast-Freeze® Flasks for FreeZone® Freeze Dry Systems are designed to be easier to handle, faster to load, and more convenient to use in the laboratory for freeze drying.

Fast-Freeze® flasks simplify lyophilization procedures by eliminating the risk of contamination from vacuum grease and reducing spillage of valuable samples. No need for washers, gaskets, retainers, or other small, easily-misplaced components. Flasks consist of a borosilicate glass bottom and silicone rubber top. Uniform glass walls maximize heat transfer while maintaining structural strength. Wide mouth design provides easy access to entire inner diameter of glassware.

  • Compatible with all major brands of freeze drying equipment, including Labconco and Virtis™

Fast-Freeze® Flasks for FreeZone® Freeze Dry Systems are designed to be easier to handle, faster to load, and more convenient to use in the laboratory for freeze drying.

Fast-Freeze® Flasks simplify lyophilization procedures with a wide-mouth design that allows easy access to samples

Fast-Freeze® Flasks simplify lyophilization procedures with a wide-mouth design that allows easy access to samples

Flat bottom, high-strength borosilicate glass flask withstands high heat and deep vacuum. Uniform glass walls maximize heat transfer while maintaining structural strength. Wide-mouth design provides full access to entire inner diameter of glassware for easy loading and cleaning. Silicone rubber snap-on tops provide reliable high-vacuum seals. Flasks should be filled no more than one-third of the volume so a maximum surface area is achieved and efficient lyophilization is assured.

Flask adapters attach to Fast-Freeze® Flasks for connection to valve ports on drying chambers or manifolds for freeze dry systems. Choose borosilicate glass or stainless steel, straight or 45° angle adapters in 0.63 cm (1/4") and 1.9 cm (3/4") diameters.

Flask Holders help to prevent sample melt back. They allow a small Fast-Freeze Flask Bottom containing the sample to be placed inside a larger Fast-Freeze Flask so that the vacuum surrounding the sample insulates it from the room atmosphere. Stainless steel. Flasks not included.

Ordering information: Each complete flask is supplied with 100 filter papers of 2.5 cm (1") diameter. Adapters sold separately. Flask bottoms, tops and filter paper are also available individually. For other glassware including Ampules, Serum Bottles, and Lyph-Lock® Flasks, contact your Avantor representative.
Customers Who Bought This Also Bought