Repeater® E3 and E3x Pipettors, Eppendorf®
Catalog #: CA75796-408
Supplier: Eppendorf
Specifications
- Description:Repeater® E3 Bundle, Including Charger Stand
- Cat. no.:CA75796-408
- Supplier No.:4987000398
Specifications
About this item
The Repeater® E3/E3x Pipettors, in combination with the Combitips Advanced® pipette tips, creates a versatile dispensing system that functions according to the positive displacement principle.
- Automatic Combitip® recognition
- Ergonomic one-button tip ejector
- Precisely programmable aspiration and dispensing speeds
- Repetitive dispensing modes
- Wide dispensing range: 1 μl to 50 ml, in 100-nanoliter increments
- Safe and ergonomic operation with 9 different selectable languages
These devices provide precise and accurate dispensing of viscous solutions (e.g. glycerol) or liquids with high vapor pressure (e.g. ethanol). Contamination-free dispensing for toxic, radioactive, and other hazardous liquids (all available modes). Large illuminated color display reduces eye strain when reading. Motor-driven function reduces human error to a minimum, and reduces the risk of repetitive strain injuries (RSI).
It is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results.
Customers Who Bought This Also Bought