Hydranal™ Water Standards for Karl Fischer Titration, Honeywell Research Chemicals
Catalog #: BJ34847-40ML
Supplier: Honeywell Research Chemicals
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Ampoule
- Description:HYDRANAL™-Water Standard 0.1, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), exact value on report of analysis
- Size:6×10×4 ml
- Cat. no.:BJ34847-40ML
- Supplier No.:34847-6X40ML
Specifications
About this item
Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application.
- Manufactured according to ISO 9001 requirements
- Report of Analysis showing the exact water content and expanded uncertainty
- Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
- Long shelf life (up to five years)
- Convenient packaging
Delivery: Supplied with detailed instruction for use.
Caution: The recommended storage temperature for product HYDRANAL™ Water Standard is 2-8°C. The product can be transported at normal ambient temperatures. In the long term, temperatures > 20°C (if possible > 8 °C) should be avoided.
Customers Who Bought This Also Bought