Column Cogent Diamond Hydride hplc, Pore:100 armstrong, Particle size: 4um, Dimension: 150MMX30MM
Catalog #: 75839-780
Supplier: MicroSolv Technology Corp
Column Cogent Diamond Hydride hplc, Pore:100 armstrong, Particle size: 4um, Dimension: 150MMX30MM
Catalog #: 75839-780
Supplier: MicroSolv Technology Corp
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
132,38 $
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
99,27 $ to 725,06 $
SureSTART™ EPA Certified Screw Vial and Cap Kits, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
161,56 $ to 173,63 $
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
28,24 $
SureSTART™ High Recovery Glass Snap Top Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
1 472,13 $ to 1 472,13 $
General Purpose Septa for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
161,06 $
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
184,38 $