GsBP-FAMEWax Glass 20Mx0.18MMx0.18UM
Catalog #: 75839-220
Supplier: General Separation Technologies, Inc.
GsBP-FAMEWax Glass 20Mx0.18MMx0.18UM
Catalog #: 75839-220
Supplier: General Separation Technologies, Inc.
Customers Who Bought This Also Bought
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
131,41 $ to 136,31 $
SureSTART™ Screw Caps, 9 mm, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
2 443,19 $ to 2 443,19 $
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Supplier: Thermo Scientific
79,21 $ to 82,14 $
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
182,14 $
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
474,54 $