GSBP-AL2O3 KCL 50Mx0.53MMx15UM
Catalog #: 75839-126
Supplier: General Separation Technologies, Inc.
Customers Who Bought This Also Bought
WHEATON® MicroLiter Screw-Thread Vials, Component Kits, Assembled, 9 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
106,65 $
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
176,80 $
Crimp Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
31,12 $
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Supplier: Thermo Scientific
3 706,93 $ to 3 706,93 $
SureSTART™ Glass Screw Top Vials, 2 ml, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
31,00 $
VWR® 11 mm Crimp Top Vials, Kits, and Inserts, Standard Opening
Supplier: VWR International
40,01 $ to 399,86 $
Plastic Vials, Caps and Convenience Kits for LCMS and HPLC
Supplier: MicroSolv Technology Corp
57,43 $
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
123,78 $