ContactUsLinkComponent
Petri Dish 100x15mm
Petri Dish, Hardy Diagnostics
Catalog #: 75838-866
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
Petri Dish 100x15mm
Petri Dish, Hardy Diagnostics
Catalog #: 75838-866
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Petri Dish
  • Size:
    15 x 100 mm
  • Cat. no.:
    75838-866
  • Supplier No.:
    330120
Specifications
About this item
Distortion-free petri dishes are optically clear.
  • Heavy Duty - reduces breakage during shipping
  • Stacking rings - guards against slippage when moving stacked plates
Hardy offers a full selection of petri plates: Mono plate Space Saver Plate Bi-Plate Tri-Plate Quad Plate Membrane Filter Methods Contacts Plate Optically clear polystyrene - for distortion free viewing. Heavy Duty - reduces breakage during shipping. Stacking rings - guards against slippage when moving stacked plates.
Customers Who Bought This Also Bought