QuEChERS Performance Standard C, Restek
Catalog #: 75834-630
Supplier: Restek
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:300 µg/ml each in acetonitrile:acetic acid (99.9:0.1)
- Description:QuEChERS Performance Standard C
- Size:1 ml/Ampule
- Cat. no.:75834-630
- Supplier No.:31155
Specifications
About this item
- Designed for use in all QuEChERS methods for pesticides in fruits and vegetables, including the original unbuffered method, AOAC 2007.01, and EN15662
- Ideal for initial method evaluations and ongoing method performance validations.
- Optimized blend of chemically compatible analytes for maximum stability and shelf life.
- Precise formulation improves data quality and operational efficiency; spend more time running samples and less time sourcing and preparing standards
- Quantitatively analyzed to confirm the composition and stability of each mixture
- Produced and tested in accordance with ISO Guide 34 and 17025 accreditation.
Certifications: AOAC 2007.01 and EN15662. Produced and tested in accordance with ISO 17034 and 17025 accreditation.
Customers Who Bought This Also Bought