Order Entry
ContactUsLinkComponent
Standard Ethylene Oxide 1ml
Ethylene Oxide, Restek
Catalog #: 75834-514
Supplier:  Restek
CAS Number:  
Ethylene Oxide, Restek
Catalog #: 75834-514
Supplier:  Restek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    50 mg/ml in methylene chloride
  • CAS No.:
    75-21-8
  • Description:
    Ethylene oxide
  • Size:
    1 ml/Ampule
  • Formula:
    C₂H₄O
  • Description:
    Ethylene oxide
  • Cat. no.:
    75834-514
  • Supplier No.:
    30620

Specifications

About this item

The test for ethylene oxide is specified in many individual drug monographs of USP24/NF19. The limit test concentration is currently 10 ppm. While the specific test solution and method will vary depending on the particular drug monograph, the solution below is suitable for most tests.

Ordering information: Ethylene oxide is available in other solvents and concentrations. Request your custom formulation at www.restek.com/solutions
Customers Who Bought This Also Bought