Order Entry
ContactUsLinkComponent
DNA Ladder 100 bp 50UG/500UL
100 bp DNA Ladder, IBI Scientific
Catalog #: 75832-464
Supplier:  IBI Scientific
CAS Number:  
DNA Ladder 100 bp 50UG/500UL
100 bp DNA Ladder, IBI Scientific
Catalog #: 75832-464
Supplier:  IBI Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    0.1 µg /µl
  • Description:
    IBI 100 bp DNA Ladder
  • Size:
    50 µg / 500 µL
  • Storage conditions:
    25°C (6 months), 4°C (12 months), -20°C (24 months)
  • Loading volume:
    0.5 µg (5µl)
  • Shipping temperature:
    Approved for shipping at room temperature or dry ice
  • Application:
    100 gel lanes
  • Intensity:
    500 bp
  • Supplier No.:
    IB01301
  • Details:
    contains 12 linear double-stranded DNA fragments

Specifications

About this item

Ready-to-use 100 bp DNA ladder with excellent resolution of 12 different sized fragments at a great value.

  • Ready-to-use DNA Ladder (pre-mixed with loading dye) - no dilution necessary
  • Concentration: 100 µg / 1 ml; (Recommended loading: 0.5 µg or 5 µl)
  • 12 fragments suitable for sizing double-stranded DNA from 100 bp to 3 Kb with enhanced intensity at 500 bp
  • Components: 100 bp DNA Ladder (pre-mixed with loading dye) and 6X DNA Loading Dye for sample loading
  • Tracking dyes: Bromophenol blue, xylene cyanol FF
  • Gel Compatibility: Agarose gels

The IBI 100bp DNA Ladder, containing 12 linear double-stranded DNA fragments, is suitable for sizing double-stranded DNA from 100bp to 3,000bp

This product comes pre-mixed with loading dye and is ready to use, no dilution necessary. There is also an extra vial of 6X loading dye included in the 50µg/500µl ladder pack.

Typical load per well is 5 µl. Product is sufficient for 100 gel lanes (applications).

The 100bp DNA Ladder is a unique combination of a number of proprietary plasmids digested with appropriate restriction enzymes and PCR products to yield 12 fragments, suitable for use as molecular weight standards for electrophoresis. The enhanced intensity of the 500bp fragment was designed for quick and easy determination of electrophoresis results.

Customers Who Bought This Also Bought