Order Entry
ContactUsLinkComponent
Mag-Bind* FFPE DNA Kit 4x96preps
Mag-Bind® Plant DNA DS 96 Kit, Omega
Catalog #: CA75832-932
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind* FFPE DNA Kit 4x96preps
Mag-Bind® Plant DNA DS 96 Kit, Omega
Catalog #: CA75832-932
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Description:
    Mag-Bind® Plant DNA DS 96 Kit
  • Size:
    4 x 96 Preps
  • Description:
    Mag-Bind® Plant DNA DS 96 Kit
  • Cat. no.:
    CA75832-932
  • Supplier No.:
    M1130-01

Specifications

About this item

The E-Z 96 Plant DNA DS Kit is designed for efficient recovery of genomic DNA up to 30 kb in size from fresh, frozen, or dried plant tissue samples rich in polysaccharides, polyphenols, or those having a lower DNA content.

  • Up to 50 mg wet tissue can be processed in less than one hour
  • Increase yields and provide high-quality DNA
  • Eliminates the need for chloroform extractions

The Mag-Bind® Plant DNA DS 96 Kit allows rapid and reliable isolation of high-quality genomic DNA from plants and other tissues that are particularly difficult to lyse or very high in polysaccharide content. The lysis and binding buffers are specifically designed to minimize co-purification of polysaccharides and polyphenols. Up 96 samples of 50 mg wet tissue (or 15 mg dry tissue) can be processed in parallel in less than one hour. The system combines CTAB-based lysis, which eliminates the need for organic solvents, with the convenience of Mag-Bind® Particles to eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates. This kit is designed for manual or fully automated high throughput preparation of genomic, chloroplast, and mitochondrial DNA. Purified DNA is suitable for PCR, restriction digestion, next-generation sequencing, and hybridization applications. There are no organic extractions thereby reducing consumables and decreasing hands-on time to allow multiple samples to be processed in parallel.

Protocols are available for the following automated platforms:
Hamilton Microlab® STAR
Hamilton Microlab® NIMBUS
KingFisher™, BioSprint®, and MagMAX® 96

Customers Who Bought This Also Bought