Specifications
- Packaging:Nalgene bottles
- Size:1x6 L
- Cat. no.:75832-012
- Supplier No.:BSS-PBS-1X6
Specifications
About this item
Cytiva manufactures PBS utilizing USP raw materials formulated in USP Water for Injection quality water.
- GMP manufactured
- PH: 7.1 -7.3 (USP <791>)
- Osmolality: 280 -320 mOsm/kg (USP <785>)
Phosphate-buffered saline (PBS) 1X is a physiological salt solution commonly found in many cell culture applications. PBS is isotonic and nontoxic to cells. Cytiva offers PBS that is sterile with endotoxin levels tested for each lot.