Sapphire 96-Well PCR Microplates, Polypropylene, Greiner Bio-One
Catalog #: 75830-560
Supplier: Greiner Bio-One
Specifications
- Skirt:Non-skirted
- Well Color:Natural
- Well Volume:0.2 ml
- Description:PCR plate
- No. of wells:96
- Cat. no.:75830-560
- Supplier No.:652201
Specifications
About this item
96-Well PCR Microplates are manufactured out of medical-grade polypropylene and are compatible with a wide range of thermal cyclers.
- Polypropylene construction
- Free of RNase, DNase, and human DNA
- Endotoxin-free
- Suitable for real-time/quantitative PCR
Microplates are thin-walled for efficient heat transfer during amplification. Packaging is labeled with product symbols for easier identification and warehousing.
Customers Who Bought This Also Bought