Septa Only, GPI 13-425, Red PTFE/Silicone, 13mm x 0.075 inch, for Screw Thread Closures
Catalog #: 75820-966
Supplier: Chemglass
Septa Only, GPI 13-425, Red PTFE/Silicone, 13mm x 0.075 inch, for Screw Thread Closures
Catalog #: 75820-966
Supplier: Chemglass
Customers Who Bought This Also Bought
Exel International Brand Quality Luer Lock Syringes, Air-Tite Products
Supplier: Air-Tite
$44.03 to $212.11
Nitric acid 67 - 70%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Supplier: BDH
$729.52
Potassium chloride 99.0-100.5% (by argentometric titration), crystals, Ultrapure Bioreagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$143.48