Poly Crimp* Seal Blue 11mm
Catalog #: 75820-946
Supplier: Chemglass
Customers Who Bought This Also Bought
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$149.89 to $566.91
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$146.85 to $576.51
Sodium chloride 0.9% (w/v) in aqueous solution isotonic saline, suitable for use as blood bank saline
Supplier: Ricca Chemical
$333.21
Conductivity Standards, Potassium Chloride (KCl) Solutions, 10 to 500 µS/cm, Thermo Scientific
Supplier: Thermo Scientific Chemicals
$72.00
Iscove's Modified Dulbecco's Medium (IMDM), Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
$105.01
L(+)-Potassium sodium tartrate tetrahydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
$961.66