Poly Crimp* Seal Blue 11mm
Catalog #: 75820-924
Supplier: Chemglass
Customers Who Bought This Also Bought
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
$77.41 to $1,124.20
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
$214.10 to $1,039.32
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
$971.75 to $3,463.97
Sodium chloride 0.9% (w/v) in aqueous solution isotonic saline, suitable for use as blood bank saline
Supplier: Ricca Chemical
$75.11