Poly Crimp* Seal Clear 11mm
Catalog #: 75820-912
Supplier: Chemglass
Customers Who Bought This Also Bought
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
$386.44 to $1,545.78
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
$2,423.69
Magnesium chloride hexahydrate 99.0-102.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
$137.92 to $1,374.20
Ocean Pacific® Ninja™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Supplier: Medicom
$49.54 to $495.19
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Dichloromethane, anhydrous ≥99.8% stabilized, DriSolv®, Supelco®
Supplier: MilliporeSigma
$283.76 to $1,668.92