Snap Plug, Clear, Conical, Material: Polyethylene, With Starburst, Size: 12mm
Catalog #: 75820-906
Supplier: Chemglass
Snap Plug, Clear, Conical, Material: Polyethylene, With Starburst, Size: 12mm
Catalog #: 75820-906
Supplier: Chemglass
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$194.14 to $694.12